Tisk…

3-Bromo-5-fluoro-4-methylpyridine 95%

Dodavatel: Apollo Scientific

PC201059-1G PC201059-5G PC201059-250MG
APOSPC201059-1GEA 3020 CZK
APOSPC201059-1G APOSPC201059-5G APOSPC201059-250MG
3-Bromo-5-fluoro-4-methylpyridine 95%
3-Bromo-5-fluoro-4-methylpyridine
Vzorec: C₆H₅BrFN
MW: 190.0130 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257616
CAS číslo: 1211517-76-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR