Tisk…

Ethyl-2-amino-4-fluorobenzoate 98%

Dodavatel: Apollo Scientific

PC201039-10G PC201039-1G PC201039-5G
APOSPC201039-10GEA 13030 CZK
APOSPC201039-10G APOSPC201039-1G APOSPC201039-5G
Ethyl-2-amino-4-fluorobenzoate 98%
Ethyl-2-amino-4-fluorobenzoate
Vzorec: C₉H₁₀FNO₂
Storage Temperature: Ambient
MDL Number: MFCD08275015
CAS číslo: 117324-05-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR