Tisk…

1-Chloro-2,4-difluoro-3-nitrobenzene 97%

Dodavatel: Apollo Scientific
Varování

PC201032-1G PC201032-250MG PC201032-5G
APOSPC201032-1GEA 1450 CZK
APOSPC201032-1G APOSPC201032-250MG APOSPC201032-5G
1-Chloro-2,4-difluoro-3-nitrobenzene 97%
1-Chloro-2,4-difluoro-3-nitrobenzene
Vzorec: C₆H₂ClF₂NO₂
MW: 193,54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10686546
CAS číslo: 1151767-58-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR