4-Amino-3-chloro-5-fluorobenzonitrile 97%
Dodavatel: Apollo Scientific
PC201029-1G
PC201029-5G
PC201029-10G
APOSPC201029-1GEA
3940
CZK
APOSPC201029-1G
APOSPC201029-5G
APOSPC201029-10G
4-Amino-3-chloro-5-fluorobenzonitrile 97%
4-Amino-3-chloro-5-fluorobenzonitrile
Vzorec:
C₇H₄ClFN₂ MW: 170,57 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18426013 CAS číslo: 1147558-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...