4,6-Dichloro-5-fluoro-2-methylpyrimidine 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC201014-1G
PC201014-250MG
APOSPC201014-1GEA
2280
CZK
APOSPC201014-1G
APOSPC201014-250MG
4,6-Dichloro-5-fluoro-2-methylpyrimidine 97%
4,6-Dichloro-5-fluoro-2-methylpyrimidine
Vzorec:
C₅H₃Cl₂FN₂ MW: 181,0000 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11520450 CAS číslo: 105806-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...