Tisk…

4-Chloro-7-fluoro-2-methylquinoline

Dodavatel: Apollo Scientific
Varování

PC200601-1G PC200601-5G
APOSPC200601-1GEA 5460 CZK
APOSPC200601-1G APOSPC200601-5G
4-Chloro-7-fluoro-2-methylquinoline
4-Chloro-7-fluoro-2-methylquinoline
Vzorec: C₁₀H₇ClFN
MW: 195,63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272333
CAS číslo: 18529-04-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR