Tisk…

2-(4-Fluorophenoxy)propionic acid 95%

Dodavatel: Apollo Scientific

PC200570-1G PC200570-5G
APOSPC200570-1GEA 2240 CZK
APOSPC200570-1G APOSPC200570-5G
2-(4-Fluorophenoxy)propionic acid 95%
2-(4-Fluorophenoxy)propionic acid
Vzorec: C₉H₉FO₃
Bod tání: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD02295725
CAS číslo: 2967-70-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR