4-Chloro-8-fluoro-2-methylquinoline
Dodavatel: Apollo Scientific
PC200565-1G
PC200565-5G
APOSPC200565-1GEA
5720
CZK
APOSPC200565-1G
APOSPC200565-5G
4-Chloro-8-fluoro-2-methylquinoline
4-Chloro-8-fluoro-2-methylquinoline
Vzorec:
C₁₀H₇ClFN Storage Temperature: Ambient |
MDL Number:
MFCD00272335 CAS číslo: 18615-59-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...