Tisk…

(3-Fluorophenoxy)acetic acid

Dodavatel: Apollo Scientific

Synonyms: 2-(3-Fluorophenoxy)acetic acid

PC200534-10G PC200534-1G PC200534-5G
APOSPC200534-10GEA 7050 CZK
APOSPC200534-10G APOSPC200534-1G APOSPC200534-5G
(3-Fluorophenoxy)acetic acid
(3-Fluorophenoxy)acetic acid
Vzorec: C₈H₇FO₃
MW: 170,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02295721
CAS číslo: 404-98-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR