3-Bromo-2,4-difluorobenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC200493-1G
PC200493-5G
PC200493-25G
APOSPC200493-1GEA
2530
CZK
APOSPC200493-1G
APOSPC200493-5G
APOSPC200493-25G
3-Bromo-2,4-difluorobenzoic acid
3-Bromo-2,4-difluorobenzoic acid
Vzorec:
C₇H₃BrF₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD09261257 CAS číslo: 651026-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...