Methyl-4-bromo-2-fluorobenzoate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 4-bromo-2-fluorobenzoate
PC200410-100G
APOSPC200410-100GEA
2780
CZK
APOSPC200410-100G
Methyl-4-bromo-2-fluorobenzoate
Methyl-4-bromo-2-fluorobenzoate
Vzorec:
C₈H₆BrFO₂ MW: 233,04 g/mol Bod tání: 60…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD06203731 CAS číslo: 179232-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...