Potassium (4-pyridyl)trifluoroborate
Dodavatel: Apollo Scientific
Synonyms:
Potassium 4-Pyridyltrifluoroborate
PC200374-5G
APOSPC200374-5GEA
3610
CZK
APOSPC200374-5G
Potassium (4-pyridyl)trifluoroborate
Potassium (4-pyridyl)trifluoroborate
Vzorec:
C₅H₄BF₃KN MW: 184,9965 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09038543 CAS číslo: 1111732-87-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...