6-Bromo-4-fluoro-1H-indazole 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC200355-1G
PC200355-250MG
PC200355-5G
APOSPC200355-1GEA
953
CZK
APOSPC200355-1G
APOSPC200355-250MG
APOSPC200355-5G
6-Bromo-4-fluoro-1H-indazole 97%
6-Bromo-4-fluoro-1H-indazole
Vzorec:
C₇H₄BrFN₂ MW: 215,0200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07781444 CAS číslo: 885520-23-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...