Tisk…

2-Chloro-6-isopropyl-3-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyridine

Dodavatel: Apollo Scientific

PC200306-1G PC200306-500MG
APOSPC200306-1GEA 16800 CZK
APOSPC200306-1G APOSPC200306-500MG
2-Chloro-6-isopropyl-3-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyridine
2-Chloro-6-isopropyl-3-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyridine
Vzorec: C₁₁H₉ClF₃N₃O
Storage Temperature: Ambient
MDL Number: MFCD14581653
CAS číslo: 1203897-93-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR