Tisk…

tert-Butyl-5-cyano-3-formyl-6-(trifluoromethyl)-1H-indole-1-carboxylate

Dodavatel: Apollo Scientific

Synonyms: 3-Formyl-6-(trifluoromethyl)-1H-indole-5-carbonitrile, N-BOC protected

PC200299-1G PC200299-500MG
APOSPC200299-1GEA 17820 CZK
APOSPC200299-1G APOSPC200299-500MG
tert-Butyl-5-cyano-3-formyl-6-(trifluoromethyl)-1H-indole-1-carboxylate
tert-Butyl-5-cyano-3-formyl-6-(trifluoromethyl)-1H-indole-1-carboxylate
Vzorec: C₁₆H₁₃F₃N₂O₃
MW: 338,29 g/mol
Bod tání: 303…304 °C
Storage Temperature: Ambient
MDL Number: MFCD20921578
CAS číslo: 1353877-93-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR