Tisk…

Methyl-3-chloro-2-piperazino-5-(trifluoromethyl)isonicotinate

Dodavatel: Apollo Scientific

PC200298-1G PC200298-500MG
APOSPC200298-1GEA 30290 CZK
APOSPC200298-1G APOSPC200298-500MG
Methyl-3-chloro-2-piperazino-5-(trifluoromethyl)isonicotinate
Methyl-3-chloro-2-piperazino-5-(trifluoromethyl)isonicotinate
Vzorec: C₁₂H₁₃ClF₃N₃O₂
MW: 323,7 g/mol
Bod tání: 75…76 °C
Storage Temperature: Ambient
MDL Number: MFCD12025894
CAS číslo: 1147979-45-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR