Tisk…

2-Chloro-6-phenyl-3-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyridine

Dodavatel: Apollo Scientific

PC200296-1G PC200296-500MG
APOSPC200296-1GEA 16800 CZK
APOSPC200296-1G APOSPC200296-500MG
2-Chloro-6-phenyl-3-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyridine
2-Chloro-6-phenyl-3-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyridine
Vzorec: C₁₄H₇ClF₃N₃O
Bod tání: 134…135 °C
Storage Temperature: Ambient
MDL Number: MFCD14581650
CAS číslo: 1198475-48-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR