Tisk…

Methyl-4-[(E)-[(2-chloropyridin-3-yl)methylidene]amino]-3-fluorobenzoate

Dodavatel: Apollo Scientific

PC200281-1G PC200281-500MG
APOSPC200281-1GEA 13430 CZK
APOSPC200281-1G APOSPC200281-500MG
Methyl-4-[(E)-[(2-chloropyridin-3-yl)methylidene]amino]-3-fluorobenzoate
Methyl-4-[(E)-[(2-chloropyridin-3-yl)methylidene]amino]-3-fluorobenzoate
Vzorec: C₁₄H₁₀ClFN₂O₂
Bod tání: 132…133 °C
Storage Temperature: Ambient
MDL Number: MFCD18904318
CAS číslo: 1296225-23-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR