Tisk…

4-(1-Methylhydrazino)-2-(trifluoromethyl)quinazoline

Dodavatel: Apollo Scientific

PC200279-1G PC200279-500MG
APOSPC200279-1GEA 14780 CZK
APOSPC200279-1G APOSPC200279-500MG
4-(1-Methylhydrazino)-2-(trifluoromethyl)quinazoline
4-(1-Methylhydrazino)-2-(trifluoromethyl)quinazoline
Vzorec: C₁₀H₉F₃N₄
Bod tání: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00793700
CAS číslo: 303148-88-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR