Tisk…

4-Piperazino-2-(2-pyridinyl)-6-(trifluoromethyl)pyrimidine

Dodavatel: Apollo Scientific

PC200269-1G PC200269-500MG
APOSPC200269-1GEA 10350 CZK
APOSPC200269-1G APOSPC200269-500MG
4-Piperazino-2-(2-pyridinyl)-6-(trifluoromethyl)pyrimidine
4-Piperazino-2-(2-pyridinyl)-6-(trifluoromethyl)pyrimidine
Vzorec: C₁₄H₁₄F₃N₅
MW: 309,3 g/mol
Bod tání: 133…134 °C
Storage Temperature: Ambient
MDL Number: MFCD11100243
CAS číslo: 1048913-94-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR