Tisk…

3-Bromo-6-(trifluoromethyl)-1H-indole-5-carbonitrile

Dodavatel: Apollo Scientific

PC200258-1G PC200258-500MG
APOSPC200258-1GEA 14780 CZK
APOSPC200258-1G APOSPC200258-500MG
3-Bromo-6-(trifluoromethyl)-1H-indole-5-carbonitrile
3-Bromo-6-(trifluoromethyl)-1H-indole-5-carbonitrile
Vzorec: C₁₀H₄BrF₃N₂
Bod tání: 214…216 °C
Storage Temperature: Ambient
MDL Number: MFCD12827794
CAS číslo: 1186404-83-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR