Tisk…

1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-pyrrolidinol

Dodavatel: Apollo Scientific

PC200255-1G PC200255-500MG
APOSPC200255-1GEA 7050 CZK
APOSPC200255-1G APOSPC200255-500MG
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-pyrrolidinol
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-pyrrolidinol
Vzorec: C₁₀H₁₀ClF₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD13561836
CAS číslo: 1219980-80-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR