Tisk…

4-(Trifluoromethyl)pyridine-2-carbaldehyde

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-(Trifluoromethyl)picolinaldehyde

PC200253-1G PC200253-500MG PC200253-100MG
APOSPC200253-1GEA 25460 CZK
APOSPC200253-1G APOSPC200253-500MG APOSPC200253-100MG
4-(Trifluoromethyl)pyridine-2-carbaldehyde
4-(Trifluoromethyl)pyridine-2-carbaldehyde
Vzorec: C₇H₄F₃NO
Storage Temperature: Refrigerator
MDL Number: MFCD11848102
CAS číslo: 132470-83-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR