Tisk…

5-Bromo-6-cyclopropyl-2-hydroxy-4-(trifluoromethyl)nicotinonitrile

Dodavatel: Apollo Scientific

PC200245-1G PC200245-500MG
APOSPC200245-1GEA 11810 CZK
APOSPC200245-1G APOSPC200245-500MG
5-Bromo-6-cyclopropyl-2-hydroxy-4-(trifluoromethyl)nicotinonitrile
5-Bromo-6-cyclopropyl-2-hydroxy-4-(trifluoromethyl)nicotinonitrile
Vzorec: C₁₀H₆BrF₃N₂O
Bod tání: 219…220 °C
Storage Temperature: Ambient
MDL Number: MFCD11553027
CAS číslo: 1092352-29-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR