Tisk…

1-(5-Chloro-2-pyridinyl)-3-(trifluoromethyl)-1H-pyrazol-5-ol

Dodavatel: Apollo Scientific

PC200244-1G PC200244-500MG PC200244-5G
APOSPC200244-1GEA 7050 CZK
APOSPC200244-1G APOSPC200244-500MG APOSPC200244-5G
1-(5-Chloro-2-pyridinyl)-3-(trifluoromethyl)-1H-pyrazol-5-ol
1-(5-Chloro-2-pyridinyl)-3-(trifluoromethyl)-1H-pyrazol-5-ol
Vzorec: C₉H₅ClF₃N₃O
MW: 263.61 g/mol
Bod tání: 60…62 °C
Storage Temperature: Ambient
MDL Number: MFCD11100238
CAS číslo: 189005-07-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR