Tisk…

5-Amino-1-[3-(trifluoromethyl)pyridin-2-yl]-1H-pyrazole-4-carbonitrile

Dodavatel: Apollo Scientific

PC200234-1G PC200234-500MG PC200234-5G
APOSPC200234-1GEA 5390 CZK
APOSPC200234-1G APOSPC200234-500MG APOSPC200234-5G
5-Amino-1-[3-(trifluoromethyl)pyridin-2-yl]-1H-pyrazole-4-carbonitrile
5-Amino-1-[3-(trifluoromethyl)pyridin-2-yl]-1H-pyrazole-4-carbonitrile
Vzorec: C₁₀H₆F₃N₅
Bod tání: 160…161 °C
Storage Temperature: Ambient
MDL Number: MFCD06808548
CAS číslo: 1019010-69-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR