Tisk…

6-(Trifluoromethyl)-1H-indole-5-carbonitrile

Dodavatel: Apollo Scientific

PC200225-1G PC200225-500MG
APOSPC200225-1GEA 19110 CZK
APOSPC200225-1G APOSPC200225-500MG
6-(Trifluoromethyl)-1H-indole-5-carbonitrile
6-(Trifluoromethyl)-1H-indole-5-carbonitrile
Vzorec: C₁₀H₅F₃N₂
Bod tání: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD12827791
CAS číslo: 1186404-61-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR