4-(Trifluoromethyl)-1H,2H,3H-pyrazolo[3,4-b]pyridin-3-one
Dodavatel: Apollo Scientific
PC200223-1G
PC200223-500MG
APOSPC200223-1GEA
11810
CZK
APOSPC200223-1G
APOSPC200223-500MG
4-(Trifluoromethyl)-1H,2H,3H-pyrazolo[3,4-b]pyridin-3-one
4-(Trifluoromethyl)-1H,2H,3H-pyrazolo[3,4-b]pyridin-3-one
Vzorec:
C₇H₄F₃N₃O MW: 203,12 g/mol Bod tání: 294…295 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD16890137 CAS číslo: 1242267-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...