Tisk…

Ethyl-3-nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95%

Dodavatel: Apollo Scientific

PC200210-1G PC200210-500MG
APOSPC200210-1GEA 15310 CZK
APOSPC200210-1G APOSPC200210-500MG
Ethyl-3-nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95%
Ethyl-3-nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Vzorec: C₁₁H₈F₃N₃O₄
Bod tání: 134…135 °C
Storage Temperature: Ambient
MDL Number: MFCD12498712
CAS číslo: 1171919-07-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR