Tisk…

3-Iodo-5-(trifluoromethyl)-2-pyridinylamine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Iodo-5-(trifluoromethyl)pyridin-2-amine, 2-Amino-3-iodo-5-(trifluoromethyl)pyridine

PC200189-5G
APOSPC200189-5GEA 5100 CZK
APOSPC200189-5G
3-Iodo-5-(trifluoromethyl)-2-pyridinylamine
3-Iodo-5-(trifluoromethyl)-2-pyridinylamine
Vzorec: C₆H₄F₃IN₂
MW: 288,01 g/mol
Bod tání: 97…98 °C
Storage Temperature: Ambient
MDL Number: MFCD10697686
CAS číslo: 211308-82-6
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR