Potassium (1-naphthalene)trifluoroborate
Dodavatel: Apollo Scientific
Synonyms:
Potassium trifluoro(naphthalen-1-yl)borate, (1-Naphthalene)trifluoroboric acid potassium salt
PC200179-10G
PC200179-1G
PC200179-5G
APOSPC200179-10GEA
7280
CZK
APOSPC200179-10G
APOSPC200179-1G
APOSPC200179-5G
Potassium (1-naphthalene)trifluoroborate
Potassium (1-naphthalene)trifluoroborate
Vzorec:
C₁₀H₇BF₃K MW: 234.07 Storage Temperature: Ambient |
MDL Number:
MFCD06659936 CAS číslo: 166328-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...