Tisk…

Ethyl-6-iodo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate

Dodavatel: Apollo Scientific

PC200169-1G PC200169-500MG
APOSPC200169-1GEA 5990 CZK
APOSPC200169-1G APOSPC200169-500MG
Ethyl-6-iodo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Ethyl-6-iodo-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Vzorec: C₁₁H₈F₃IN₂O₂
Bod tání: 148…149 °C
Storage Temperature: Ambient
MDL Number: MFCD06496246
CAS číslo: 900019-79-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR