Tisk…

5-Bromo-2-chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile

Dodavatel: Apollo Scientific

PC200161-1G PC200161-500MG
APOSPC200161-1GEA 5990 CZK
APOSPC200161-1G APOSPC200161-500MG
5-Bromo-2-chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile
5-Bromo-2-chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile
Vzorec: C₈H₃BrClF₃N₂
MW: 299,48 g/mol
Bod tání: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD11841008
CAS číslo: 1135283-49-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR