3-(Trifluoromethyl)-5-isoxazolamine 95+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Amino-3-(trifluoromethyl)isoxazole
PC200153-1G
PC200153-500MG
APOSPC200153-1GEA
8930
CZK
APOSPC200153-1G
APOSPC200153-500MG
3-(Trifluoromethyl)-5-isoxazolamine 95+%
3-(Trifluoromethyl)-5-isoxazolamine
Vzorec:
C₄H₃F₃N₂O MW: 152,08 g/mol Bod tání: 53…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD04972407 CAS číslo: 108655-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...