Potassium-3-carboxyphenyltrifluoroborate
Dodavatel: Apollo Scientific
PC200140-10G
PC200140-1G
PC200140-5G
APOSPC200140-10GEA
9690
CZK
APOSPC200140-10G
APOSPC200140-1G
APOSPC200140-5G
Potassium-3-carboxyphenyltrifluoroborate
Potassium-3-carboxyphenyltrifluoroborate
Vzorec:
C₇H₅BF₃KO₂ MW: 228.02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07784423 CAS číslo: 850313-91-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...