Tisk…

Potassium-4-(ethoxycarbonyl)phenyltrifluoroborate

Dodavatel: Apollo Scientific

PC200138-1G PC200138-5G
APOSPC200138-1GEA 2700 CZK
APOSPC200138-1G APOSPC200138-5G
Potassium-4-(ethoxycarbonyl)phenyltrifluoroborate
Potassium-4-(ethoxycarbonyl)phenyltrifluoroborate
Vzorec: C₉H₉BF₃KO₂
Storage Temperature: Ambient
MDL Number: MFCD09800743
CAS číslo: 1439402-07-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR