Tisk…

Potassium-4-(4-morpholinylcarbonyl)phenyltrifluoroborate

Dodavatel: Apollo Scientific

Synonyms: Potassium trifluoro(4-(morpholine-4-carbonyl)phenyl)borate

PC200137-1G PC200137-500MG PC200137-5G
APOSPC200137-1GEA 2900 CZK
APOSPC200137-1G APOSPC200137-500MG APOSPC200137-5G
Potassium-4-(4-morpholinylcarbonyl)phenyltrifluoroborate
Potassium-4-(4-morpholinylcarbonyl)phenyltrifluoroborate
Vzorec: C₁₁H₁₂BF₃KNO₂
MW: 297,123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800742
CAS číslo: 1314957-11-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR