Tisk…

Methyl-2-chloro-5-fluoronicotinate

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 2-chloro-5-fluoronicotinate

PC200135-1G PC200135-500MG
APOSPC200135-1GEA 6020 CZK
APOSPC200135-1G APOSPC200135-500MG
Methyl-2-chloro-5-fluoronicotinate
Methyl-2-chloro-5-fluoronicotinate
Vzorec: C₇H₅ClFNO₂
MW: 189,57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09763661
CAS číslo: 847729-27-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR