Tisk…

Potassium-5-formylthiophene-2-trifluoroborate

Dodavatel: Apollo Scientific

PC200134-1G PC200134-500MG PC200134-5G
APOSPC200134-1GEA 916 CZK
APOSPC200134-1G APOSPC200134-500MG APOSPC200134-5G
Potassium-5-formylthiophene-2-trifluoroborate
Potassium-5-formylthiophene-2-trifluoroborate
Vzorec: C₅H₃BF₃KOS
Storage Temperature: Ambient
MDL Number: MFCD09800740
CAS číslo: 1025113-78-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR