Potassium-3-formylphenyltrifluoroborate
Dodavatel: Apollo Scientific
Synonyms:
Potassium trifluoro(3-formylphenyl)borate
PC200133-5G
APOSPC200133-5GEA
2110
CZK
APOSPC200133-5G
Potassium-3-formylphenyltrifluoroborate
Potassium-3-formylphenyltrifluoroborate
Vzorec:
C₇H₅BF₃KO MW: 212,02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07784378 CAS číslo: 871231-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...