Tisk…

2,4-Dichloro-3-(trifluoromethyl)pyridine

Dodavatel: Apollo Scientific
Varování

PC200112-1G PC200112-500MG PC200112-5G
APOSPC200112-1GEA 6020 CZK
APOSPC200112-1G APOSPC200112-500MG APOSPC200112-5G
2,4-Dichloro-3-(trifluoromethyl)pyridine
2,4-Dichloro-3-(trifluoromethyl)pyridine
Vzorec: C₆H₂Cl₂F₃N
Storage Temperature: Ambient
MDL Number: MFCD12547046
CAS číslo: 1186194-98-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR