Potassium ethynyltrifluoroborate
Dodavatel: Apollo Scientific
|
Varování
|
PC200093-1G
PC200093-500MG
APOSPC200093-1GEA
2170
CZK
APOSPC200093-1G
APOSPC200093-500MG
Potassium ethynyltrifluoroborate
Potassium ethynyltrifluoroborate
Vzorec:
C₂HBF₃K Bod tání: 208…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD10700167 CAS číslo: 892869-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...