Tisk…

Ethyl-4,6-dihydroxy-2-(trifluoromethyl)nicotinate

Dodavatel: Apollo Scientific

PC200071-1G PC200071-500MG PC200071-5G
APOSPC200071-1GEA 7670 CZK
APOSPC200071-1G APOSPC200071-500MG APOSPC200071-5G
Ethyl-4,6-dihydroxy-2-(trifluoromethyl)nicotinate
Ethyl-4,6-dihydroxy-2-(trifluoromethyl)nicotinate
Vzorec: C₉H₈F₃NO₄
MW: 251,16 g/mol
Bod tání: 217…219 °C
Storage Temperature: Ambient
MDL Number: MFCD09881353
CAS číslo: 103900-77-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR