Tisk…

Ethyl (3-chloro-5-(trifluoromethyl)pyridin-2-yl)acetate

Dodavatel: Apollo Scientific
Varování

PC200061-1G PC200061-500MG PC200061-5G
APOSPC200061-1GEA 5490 CZK
APOSPC200061-1G APOSPC200061-500MG APOSPC200061-5G
Ethyl (3-chloro-5-(trifluoromethyl)pyridin-2-yl)acetate
Ethyl (3-chloro-5-(trifluoromethyl)pyridin-2-yl)acetate
Vzorec: C₁₀H₉ClF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD10568306
CAS číslo: 1053656-47-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR