2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]acetonitrile
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)acetonitrile
PC200054-1G
PC200054-5G
PC200054-10G
APOSPC200054-1GEA
4170
CZK
APOSPC200054-1G
APOSPC200054-5G
APOSPC200054-10G
2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]acetonitrile
2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]acetonitrile
Vzorec:
C₈H₄ClF₃N₂ Storage Temperature: Ambient |
MDL Number:
MFCD08444379 CAS číslo: 157764-10-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...