3-[3-(Trifluoromethyl)-1H-pyrazol-4-yl]-1-propanol
Dodavatel: Apollo Scientific
Synonyms:
4-(3-Hydroxyprop-1-yl)-3-(trifluoromethyl)-1H-pyrazole
PC200043-1G
PC200043-500MG
PC200043-5G
APOSPC200043-1GEA
7670
CZK
APOSPC200043-1G
APOSPC200043-500MG
APOSPC200043-5G
3-[3-(Trifluoromethyl)-1H-pyrazol-4-yl]-1-propanol
3-[3-(Trifluoromethyl)-1H-pyrazol-4-yl]-1-propanol
Vzorec:
C₇H₉F₃N₂O MW: 194,16 g/mol Bod tání: 79…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00794530 CAS číslo: 169213-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...