Tisk…

2-[(2-Fluoro-6-nitrophenyl)sulphanyl]acetic acid

Dodavatel: Apollo Scientific
Varování

PC200017-1G PC200017-500MG
APOSPC200017-1GEA 15310 CZK
APOSPC200017-1G APOSPC200017-500MG
2-[(2-Fluoro-6-nitrophenyl)sulphanyl]acetic acid
2-[(2-Fluoro-6-nitrophenyl)sulphanyl]acetic acid
Vzorec: C₈H₆FNO₄S
MW: 231.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22573544
CAS číslo: 1247021-61-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR