4-(Chloromethyl)-1-fluoro-2-iodobenzene
Dodavatel: Apollo Scientific
PC200009-1G
PC200009-500MG
APOSPC200009-1GEA
17200
CZK
APOSPC200009-1G
APOSPC200009-500MG
4-(Chloromethyl)-1-fluoro-2-iodobenzene
4-(Chloromethyl)-1-fluoro-2-iodobenzene
Vzorec:
C₇H₅ClFI Storage Temperature: Ambient |
MDL Number:
MFCD18396969 CAS číslo: 1261837-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...