3-Chloro-2-methyl-5-trifluoromethylpyridine
Dodavatel: Apollo Scientific
|
Varování
|
PC1939-1G
PC1939-5G
APOSPC1939-1GEA
2690
CZK
APOSPC1939-1G
APOSPC1939-5G
3-Chloro-2-methyl-5-trifluoromethylpyridine
3-Chloro-2-methyl-5-trifluoromethylpyridine
Vzorec:
C₇H₅ClF₃N MW: 195,57 g/mol Bod varu: 56…59 °C/15mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00068123 CAS číslo: 175277-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...