4-Chloro-6-methyl-5-nitro-2-(trifluoromethyl)quinoline 95%
Dodavatel: Apollo Scientific
PC1936M-1G
PC1936M-5G
APOSPC1936M-1GEA
744
CZK
APOSPC1936M-1G
APOSPC1936M-5G
4-Chloro-6-methyl-5-nitro-2-(trifluoromethyl)quinoline 95%
4-Chloro-6-methyl-5-nitro-2-(trifluoromethyl)quinoline
Vzorec:
C₁₁H₆ClF₃N₂O₂ MW: 290,63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00126373 CAS číslo: 175203-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...